Tisk…

5-(Hydroxymethyl)-1-methyl-3-phenyl-1H-pyrazole

Dodavatel: Apollo Scientific
Varování

OR9176-1G OR9176-250MG
APOSOR9176-1GEA 4770 CZK
APOSOR9176-1G APOSOR9176-250MG
5-(Hydroxymethyl)-1-methyl-3-phenyl-1H-pyrazole
5-(Hydroxymethyl)-1-methyl-3-phenyl-1H-pyrazole
Vzorec: C₁₁H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD08271942
CAS číslo: 864068-97-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR