Tisk…

1-Ethyl-1H-imidazole-5-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR917546-1G OR917546-250MG
APOSOR917546-1GEA 5360 CZK
APOSOR917546-1G APOSOR917546-250MG
1-Ethyl-1H-imidazole-5-carboxylic acid 95%
1-Ethyl-1H-imidazole-5-carboxylic acid
Vzorec: C₆H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08445747
CAS číslo: 71925-11-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR