N-Methyl-3-oxo-3-phenylpropanamide 95%
Dodavatel: Apollo Scientific
OR917336-1G
OR917336-250MG
APOSOR917336-1GEA
14290
CZK
APOSOR917336-1G
APOSOR917336-250MG
N-Methyl-3-oxo-3-phenylpropanamide 95%
N-Methyl-3-oxo-3-phenylpropanamide
Vzorec:
C₁₀H₁₁NO₂ MW: 177,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24549341 CAS číslo: 197852-01-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...