Tisk…

Boc-D-homoserine lactone 97%

Dodavatel: Apollo Scientific

Synonyms: Boc-D-Homoserine lactone

OR917300-5G
APOSOR917300-5GEA 2980 CZK
APOSOR917300-5G
Boc-D-homoserine lactone 97%
Boc-D-homoserine lactone
Vzorec: C₉H₁₅NO₄
MW: 201.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037376
CAS číslo: 67198-86-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR