Tisk…

3,4-Dichloro-5-nitropyridine 95%

Dodavatel: Apollo Scientific

OR917240-1G
APOSOR917240-1GEA 1490 CZK
APOSOR917240-1G
3,4-Dichloro-5-nitropyridine 95%
3,4-Dichloro-5-nitropyridine
Vzorec: C₅H₂Cl₂N₂O₂
MW: 192,988 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09910258
CAS číslo: 56809-84-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR