Tisk…

2,2-Dimethylbut-3-ynoic acid 95%

Dodavatel: Apollo Scientific

OR917188-1G
APOSOR917188-1GEA 4930 CZK
APOSOR917188-1G
2,2-Dimethylbut-3-ynoic acid 95%
2,2-Dimethylbut-3-ynoic acid
Vzorec: C₆H₈O₂
MW: 112,127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093337
CAS číslo: 56663-76-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR