Tisk…

5-Bromo-4-methylpicolinaldehyde 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-4-methylpyridine-2-carbaldehyde

OR917140-1G OR917140-250MG
APOSOR917140-1GEA 3610 CZK
APOSOR917140-1G APOSOR917140-250MG
5-Bromo-4-methylpicolinaldehyde 97%
5-Bromo-4-methylpicolinaldehyde
Vzorec: C₇H₆BrNO
MW: 200.033 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07375095
CAS číslo: 886364-94-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR