Tisk…

4-Chloro-3-hydroxy butyronitrile 95%

Dodavatel: Apollo Scientific

OR917127-1G OR917127-5G
APOSOR917127-1GEA 9100 CZK
APOSOR917127-1G APOSOR917127-5G
4-Chloro-3-hydroxy butyronitrile 95%
4-Chloro-3-hydroxy butyronitrile
Vzorec: C₄H₆ClNO
MW: 119,55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783068
CAS číslo: 105-33-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR