Tisk…

3,4-Dibromobenzoic acid 97%

Dodavatel: Apollo Scientific

OR917118-5G
APOSOR917118-5GEA 2400 CZK
APOSOR917118-5G
3,4-Dibromobenzoic acid 97%
3,4-Dibromobenzoic acid
Vzorec: C₇H₄Br₂O₂
MW: 279,913 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00094403
CAS číslo: 619-03-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR