Tisk…

(2-Oxo-1,3-benzoxazol-3(2H)-yl)acetaldehyde 95%

Dodavatel: Apollo Scientific

OR917094-1G OR917094-250MG
APOSOR917094-1GEA 11380 CZK
APOSOR917094-1G APOSOR917094-250MG
(2-Oxo-1,3-benzoxazol-3(2H)-yl)acetaldehyde 95%
(2-Oxo-1,3-benzoxazol-3(2H)-yl)acetaldehyde
Vzorec: C₉H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD09864427
CAS číslo: 13610-81-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR