Cyclohexylsulphamoyl chloride 95%
Dodavatel: Apollo Scientific
OR917027-1G
OR917027-250MG
APOSOR917027-1GEA
15110
CZK
APOSOR917027-1G
APOSOR917027-250MG
Cyclohexylsulphamoyl chloride 95%
Cyclohexylsulphamoyl chloride
Vzorec:
C₆H₁₂ClNO₂S MW: 197.683 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11109158 CAS číslo: 10314-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...