1-Methyl-1H-indazole-4-carboxylic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR916985-1G
OR916985-250MG
APOSOR916985-1GEA
4630
CZK
APOSOR916985-1G
APOSOR916985-250MG
1-Methyl-1H-indazole-4-carboxylic acid 95%
1-Methyl-1H-indazole-4-carboxylic acid
Vzorec:
C₉H₈N₂O₂ MW: 176,172 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15071442 CAS číslo: 1071433-05-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...