Tisk…

4-Chloroisoindoline hydrochloride 95%

Dodavatel: Apollo Scientific

OR916878-1G
APOSOR916878-1GEA 3380 CZK
APOSOR916878-1G
4-Chloroisoindoline hydrochloride 95%
4-Chloroisoindoline hydrochloride
Vzorec: C₈H₈ClN·ClH
MW: 190,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110370
CAS číslo: 924304-73-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR