1-(2,4-Dihydroxy-5-isopropylphenyl)ethanone 95%
Dodavatel: Apollo Scientific
OR916824-1G
OR916824-250MG
APOSOR916824-1GEA
3190
CZK
APOSOR916824-1G
APOSOR916824-250MG
1-(2,4-Dihydroxy-5-isopropylphenyl)ethanone 95%
1-(2,4-Dihydroxy-5-isopropylphenyl)ethanone
Vzorec:
C₁₁H₁₄O₃ MW: 194,227 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12827976 CAS číslo: 747414-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...