Tisk…

(4-Methyl-thiazol-2-yl)-acetic acid ethyl ester 95%

Dodavatel: Apollo Scientific

OR916804-1G OR916804-250MG
APOSOR916804-1GEA 17990 CZK
APOSOR916804-1G APOSOR916804-250MG
(4-Methyl-thiazol-2-yl)-acetic acid ethyl ester 95%
(4-Methyl-thiazol-2-yl)-acetic acid ethyl ester
Vzorec: C₈H₁₁NO₂S
Storage Temperature: Ambient
MDL Number: MFCD01819872
CAS číslo: 51221-43-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR