Tisk…

1,2,6-Trimethyl-1H-indole-3-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR916793-1G OR916793-250MG
APOSOR916793-1GEA 11380 CZK
APOSOR916793-1G APOSOR916793-250MG
1,2,6-Trimethyl-1H-indole-3-carbaldehyde 95%
1,2,6-Trimethyl-1H-indole-3-carbaldehyde
Vzorec: C₁₂H₁₃NO
MW: 187,238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06589824
CAS číslo: 842971-69-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR