Tisk…

Allyl tert-butyl carbonate 98%

Dodavatel: Apollo Scientific

OR916774-1G OR916774-25G OR916774-5G
APOSOR916774-1GEA 1700 CZK
APOSOR916774-1G APOSOR916774-25G APOSOR916774-5G
Allyl tert-butyl carbonate 98%
Allyl tert-butyl carbonate
Vzorec: C₈H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD06658296
CAS číslo: 70122-89-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR