Tisk…

1-Boc-4-(4-chlorophenyl)-4-carboxypiperidine 97%

Dodavatel: Apollo Scientific

OR916755-1G OR916755-250MG OR916755-500MG
APOSOR916755-1GEA 14950 CZK
APOSOR916755-1G APOSOR916755-250MG APOSOR916755-500MG
1-Boc-4-(4-chlorophenyl)-4-carboxypiperidine 97%
1-Boc-4-(4-chlorophenyl)-4-carboxypiperidine
Vzorec: C₁₇H₂₂ClNO₄
MW: 339.814 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05861537
CAS číslo: 644981-94-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR