4-Bromo-2-(bromomethyl)thiazole 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR916732-1G
OR916732-250MG
APOSOR916732-1GEA
18820
CZK
APOSOR916732-1G
APOSOR916732-250MG
4-Bromo-2-(bromomethyl)thiazole 97%
4-Bromo-2-(bromomethyl)thiazole
Vzorec:
C₄H₃Br₂NS MW: 256,946 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD17168686 CAS číslo: 1138333-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...