Tisk…

Methyl 4-cyano-2-methylbenzoate 95%

Dodavatel: Apollo Scientific

OR916717-5G
APOSOR916717-5GEA 2820 CZK
APOSOR916717-5G
Methyl 4-cyano-2-methylbenzoate 95%
Methyl 4-cyano-2-methylbenzoate
Vzorec: C₁₀H₉NO₂
MW: 175,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399110
CAS číslo: 103261-67-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR