Ethyl 1,2,3-thiadiazole-5-carboxylate 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR916703-1G
OR916703-500MG
OR916703-250MG
APOSOR916703-1GEA
14290
CZK
APOSOR916703-1G
APOSOR916703-500MG
APOSOR916703-250MG
Ethyl 1,2,3-thiadiazole-5-carboxylate 95%
Ethyl 1,2,3-thiadiazole-5-carboxylate
Vzorec:
C₅H₆N₂O₂S MW: 158,178 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00971933 CAS číslo: 4100-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...