Tisk…

5-Chloro-1H-pyrazole-3-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR916683-1G OR916683-250MG
APOSOR916683-1GEA 6190 CZK
APOSOR916683-1G APOSOR916683-250MG
5-Chloro-1H-pyrazole-3-carboxylic acid 95%
5-Chloro-1H-pyrazole-3-carboxylic acid
Vzorec: C₄H₃ClN₂O₂
MW: 146,532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09608056
CAS číslo: 881668-70-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR