Tisk…

3-Bromo-1-(3-chloropyridin-2-yl)-1H-pyrazole-5-carboxylic acid 96%

Dodavatel: Apollo Scientific

OR916671-25G
APOSOR916671-25GEA 1950 CZK
APOSOR916671-25G
3-Bromo-1-(3-chloropyridin-2-yl)-1H-pyrazole-5-carboxylic acid 96%
3-Bromo-1-(3-chloropyridin-2-yl)-1H-pyrazole-5-carboxylic acid
Vzorec: C₉H₅BrClN₃O₂
MW: 302,512 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689880
CAS číslo: 500011-86-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR