Tisk…

4-(Pyrrolidin-1-ylmethyl)thiophene-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR916631-1G OR916631-250MG
APOSOR916631-1GEA 11380 CZK
APOSOR916631-1G APOSOR916631-250MG
4-(Pyrrolidin-1-ylmethyl)thiophene-2-carbaldehyde 95%
4-(Pyrrolidin-1-ylmethyl)thiophene-2-carbaldehyde
Vzorec: C₁₀H₁₃NOS
MW: 195,281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06803374
CAS číslo: 893745-81-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR