Tisk…

4-Hydroxy-b-nitrostyrene 95%

Dodavatel: Apollo Scientific

Synonyms: 4-(2-Nitrovinyl)phenol

OR916627-5G
APOSOR916627-5GEA 4800 CZK
APOSOR916627-5G
4-Hydroxy-b-nitrostyrene 95%
4-Hydroxy-b-nitrostyrene
Vzorec: C₈H₇NO₃
MW: 165,146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060551
CAS číslo: 3179-08-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR