Tisk…

2,4-Dihydroxy-5-methoxybenzaldehyde 95%

Dodavatel: Apollo Scientific

OR916496-1G OR916496-250MG
APOSOR916496-1GEA 20270 CZK
APOSOR916496-1G APOSOR916496-250MG
2,4-Dihydroxy-5-methoxybenzaldehyde 95%
2,4-Dihydroxy-5-methoxybenzaldehyde
Vzorec: C₈H₈O₄
MW: 168,147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07626333
CAS číslo: 51061-83-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR