Tisk…

4-Methyl-8-hydroxyquinoline 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Methylquinolin-8-ol

OR916329-1G OR916329-250MG
APOSOR916329-1GEA 5030 CZK
APOSOR916329-1G APOSOR916329-250MG
4-Methyl-8-hydroxyquinoline 95%
4-Methyl-8-hydroxyquinoline
Vzorec: C₁₀H₉NO
MW: 159,185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02170459
CAS číslo: 3846-73-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR