Tisk…

4-(Cyclopropyl)thiazole 95%

Dodavatel: Apollo Scientific

OR916250-1G OR916250-250MG
APOSOR916250-1GEA 8200 CZK
APOSOR916250-1G APOSOR916250-250MG
4-(Cyclopropyl)thiazole 95%
4-(Cyclopropyl)thiazole
Vzorec: C₆H₇NS
Storage Temperature: Ambient
MDL Number: MFCD07390348
CAS číslo: 433217-34-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR