Tisk…

2-Methyl-2-(3-nitrophenyl)propanenitrile 95%

Dodavatel: Apollo Scientific

OR916221-10G OR916221-1G OR916221-5G
APOSOR916221-10GEA 20930 CZK
APOSOR916221-10G APOSOR916221-1G APOSOR916221-5G
2-Methyl-2-(3-nitrophenyl)propanenitrile 95%
2-Methyl-2-(3-nitrophenyl)propanenitrile
Vzorec: C₁₀H₁₀N₂O₂
MW: 190,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11036617
CAS číslo: 915394-28-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR