2-[3-(Chloromethyl)phenyl]isoindole-1,3-dione 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR915271-1G
OR915271-5G
APOSOR915271-1GEA
6450
CZK
APOSOR915271-1G
APOSOR915271-5G
2-[3-(Chloromethyl)phenyl]isoindole-1,3-dione 98%
2-[3-(Chloromethyl)phenyl]isoindole-1,3-dione
Vzorec:
C₁₅H₁₀ClNO₂ MW: 271.698 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28122691 CAS číslo: 93944-92-4 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...