Tisk…

4-Methanesulphonyl-2-methylaniline 95%

Dodavatel: Apollo Scientific

OR914708-1G OR914708-250MG OR914708-5G
APOSOR914708-1GEA 9920 CZK
APOSOR914708-1G APOSOR914708-250MG APOSOR914708-5G
4-Methanesulphonyl-2-methylaniline 95%
4-Methanesulphonyl-2-methylaniline
Vzorec: C₈H₁₁NO₂S
Storage Temperature: Ambient
MDL Number: MFCD09909541
CAS číslo: 252562-00-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR