Tisk…

2-Methyl-4-o-tolylbut-3-yn-2-ol 98%

Dodavatel: Apollo Scientific

OR914639-1G OR914639-25G OR914639-5G
APOSOR914639-1GEA 3540 CZK
APOSOR914639-1G APOSOR914639-25G APOSOR914639-5G
2-Methyl-4-o-tolylbut-3-yn-2-ol 98%
2-Methyl-4-o-tolylbut-3-yn-2-ol
Vzorec: C₁₂H₁₄O
MW: 174.239 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13195741
CAS číslo: 40888-14-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR