Tisk…

4-(Cyclopropylmethoxy)aniline hydrochloride 98%

Dodavatel: Apollo Scientific

OR914497-10G OR914497-1G OR914497-5G
APOSOR914497-10GEA 22950 CZK
APOSOR914497-10G APOSOR914497-1G APOSOR914497-5G
4-(Cyclopropylmethoxy)aniline hydrochloride 98%
4-(Cyclopropylmethoxy)aniline hydrochloride
Vzorec: C₁₀H₁₃NO·ClH
MW: 199,677 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15200817
CAS číslo: 1158573-73-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR