Tisk…

N,N-Dimethylbenzenesulphonamide 97%

Dodavatel: Apollo Scientific
Varování

OR914486-1G OR914486-5G
APOSOR914486-1GEA 4270 CZK
APOSOR914486-1G APOSOR914486-5G
N,N-Dimethylbenzenesulphonamide 97%
N,N-Dimethylbenzenesulphonamide
Vzorec: C₈H₁₁NO₂S
MW: 185,243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00514037
CAS číslo: 14417-01-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR