Tisk…

Methyl (2E)-3-(4-methoxyphenyl)prop-2-enoate 98%

Dodavatel: Apollo Scientific

Synonyms: Methyl 3-(4-methoxyphenyl)acrylate

OR914232-100G OR914232-25G OR914232-5G
APOSOR914232-100GEA 30190 CZK
APOSOR914232-100G APOSOR914232-25G APOSOR914232-5G
Methyl (2E)-3-(4-methoxyphenyl)prop-2-enoate 98%
Methyl (2E)-3-(4-methoxyphenyl)prop-2-enoate
Vzorec: C₁₁H₁₂O₃
MW: 192,211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00188501
CAS číslo: 832-01-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR