1-(2H-Chromen-3-yl)ethanone 97%
Dodavatel: Apollo Scientific
OR914081-1G
OR914081-5G
APOSOR914081-1GEA
7250
CZK
APOSOR914081-1G
APOSOR914081-5G
1-(2H-Chromen-3-yl)ethanone 97%
1-(2H-Chromen-3-yl)ethanone
Vzorec:
C₁₁H₁₀O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01662270 CAS číslo: 51593-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...