(S)-2-Methylbutane-2-sulphinamide 95%
Dodavatel: Apollo Scientific
OR913884-1G
OR913884-250MG
APOSOR913884-1GEA
12400
CZK
APOSOR913884-1G
APOSOR913884-250MG
(S)-2-Methylbutane-2-sulphinamide 95%
(S)-2-Methylbutane-2-sulphinamide
Vzorec:
C₅H₁₃NOS Storage Temperature: Ambient |
MDL Number:
MFCD11707201 CAS číslo: 861821-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...