Tisk…

tert-Butyl (3S)-3-cyanopiperazine-1-carboxylate 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-tert-Butyl 3-cyanopiperazine-1-carboxylate

OR913839-1G OR913839-250MG
APOSOR913839-1GEA 27910 CZK
APOSOR913839-1G APOSOR913839-250MG
tert-Butyl (3S)-3-cyanopiperazine-1-carboxylate 95%
tert-Butyl (3S)-3-cyanopiperazine-1-carboxylate
Vzorec: C₁₀H₁₇N₃O₂
MW: 211,261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09263269
CAS číslo: 1217650-60-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR