Tisk…

3-Methoxy-4-methylsulphonylaniline 95%

Dodavatel: Apollo Scientific
Varování

OR913835-100MG OR913835-1G OR913835-250MG
APOSOR913835-100MGEA 8070 CZK
APOSOR913835-100MG APOSOR913835-1G APOSOR913835-250MG
3-Methoxy-4-methylsulphonylaniline 95%
3-Methoxy-4-methylsulphonylaniline
Vzorec: C₈H₁₁NO₃S
MW: 201,243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13188510
CAS číslo: 75259-31-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR