(R)-1,4-Benzodioxane-2-carboxylic acid 95%
Dodavatel: Apollo Scientific
OR913611-1G
OR913611-250MG
APOSOR913611-1GEA
5060
CZK
APOSOR913611-1G
APOSOR913611-250MG
(R)-1,4-Benzodioxane-2-carboxylic acid 95%
(R)-1,4-Benzodioxane-2-carboxylic acid
Vzorec:
C₉H₈O₄ MW: 180,157 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00239407 CAS číslo: 70918-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...