1-Methyl-5-[(methylamino)methyl]-3-phenyl-1H-pyrazole 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR9136-1G
OR9136-250MG
APOSOR9136-1GEA
8440
CZK
APOSOR9136-1G
APOSOR9136-250MG
1-Methyl-5-[(methylamino)methyl]-3-phenyl-1H-pyrazole 95%
1-Methyl-5-[(methylamino)methyl]-3-phenyl-1H-pyrazole
Vzorec:
C₁₂H₁₅N₃ MW: 201,268 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08271945 CAS číslo: 864068-98-4 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...