Tisk…

(R)-(+)-Methyl p-tolyl sulphoxide 95%

Dodavatel: Apollo Scientific

Synonyms: (R)-1-Methyl-4-(methylsulphinyl)benzene

OR913597-1G OR913597-250MG
APOSOR913597-1GEA 13860 CZK
APOSOR913597-1G APOSOR913597-250MG
(R)-(+)-Methyl p-tolyl sulphoxide 95%
(R)-(+)-Methyl p-tolyl sulphoxide
Vzorec: C₈H₁₀OS
MW: 154,229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151502
CAS číslo: 1519-39-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR