Tisk…

Methyl 4-aminophenylacetate hydrochloride 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: hydrochloride, Methyl 2-(4-aminophenyl)acetate

OR913528-25G OR913528-5G
APOSOR913528-25GEA 10350 CZK
APOSOR913528-25G APOSOR913528-5G
Methyl 4-aminophenylacetate hydrochloride 98%
Methyl 4-aminophenylacetate hydrochloride
Vzorec: C₉H₁₁NO₂·ClH
MW: 201,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06796526
CAS číslo: 83528-16-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR