(R)-Methyl pyrrolidine-3-carboxylate hydrochloride 95%
Dodavatel: Apollo Scientific
OR913422-1G
OR913422-250MG
APOSOR913422-1GEA
4140
CZK
APOSOR913422-1G
APOSOR913422-250MG
(R)-Methyl pyrrolidine-3-carboxylate hydrochloride 95%
(R)-Methyl pyrrolidine-3-carboxylate hydrochloride
Vzorec:
C₆H₁₁NO₂·ClH MW: 165,618 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11840195 CAS číslo: 874964-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...