Tisk…

2-Methoxy-3-methylpyridine 95%

Dodavatel: Apollo Scientific

OR913222-1G OR913222-250MG
APOSOR913222-1GEA 3060 CZK
APOSOR913222-1G APOSOR913222-250MG
2-Methoxy-3-methylpyridine 95%
2-Methoxy-3-methylpyridine
Vzorec: C₇H₉NO
MW: 123.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367922
CAS číslo: 19230-59-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR