Tisk…

6-Methoxy-1H-indazole-3-carbonitrile 95%

Dodavatel: Apollo Scientific
Varování

OR913173-1G OR913173-250MG
APOSOR913173-1GEA 7710 CZK
APOSOR913173-1G APOSOR913173-250MG
6-Methoxy-1H-indazole-3-carbonitrile 95%
6-Methoxy-1H-indazole-3-carbonitrile
Vzorec: C₉H₇N₃O
MW: 173,171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659784
CAS číslo: 691900-59-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR