Tisk…
2-Oxo-1,2-dihydro-[1,8]naphthyridine-3-carboxylic acid 98%
Dodavatel: Apollo Scientific
Synonyms: 2-Oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylic acid
OR913168-1G OR913168-250MG
APOSOR913168-1GEA 6020 CZK
APOSOR913168-1G APOSOR913168-250MG
2-Oxo-1,2-dihydro-[1,8]naphthyridine-3-carboxylic acid 98%
2-Oxo-1,2-dihydro-[1,8]naphthyridine-3-carboxylic acid
Vzorec: C₉H₆N₂O₃
MW: 190,156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659017
CAS číslo: 5175-14-4