Tisk…

4-Hydroxy-3-methylthiophenol 97%

Dodavatel: Apollo Scientific

OR913154-1G OR913154-250MG
APOSOR913154-1GEA 19440 CZK
APOSOR913154-1G APOSOR913154-250MG
4-Hydroxy-3-methylthiophenol 97%
4-Hydroxy-3-methylthiophenol
Vzorec: C₇H₈OS
Storage Temperature: Ambient
MDL Number: MFCD06657554
CAS číslo: 32281-01-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR