4-Methoxyindole-3-carboxylic acid 96%
Dodavatel: Apollo Scientific
OR913081-1G
OR913081-250MG
OR913081-5G
APOSOR913081-1GEA
7440
CZK
APOSOR913081-1G
APOSOR913081-250MG
APOSOR913081-5G
4-Methoxyindole-3-carboxylic acid 96%
4-Methoxyindole-3-carboxylic acid
Vzorec:
C₁₀H₉NO₃ MW: 191,183 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05664011 CAS číslo: 203937-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...