7-Methoxy-1H-indole-3-carboxylic acid 95%
Dodavatel: Apollo Scientific
OR913079-1G
OR913079-250MG
APOSOR913079-1GEA
4100
CZK
APOSOR913079-1G
APOSOR913079-250MG
7-Methoxy-1H-indole-3-carboxylic acid 95%
7-Methoxy-1H-indole-3-carboxylic acid
Vzorec:
C₁₀H₉NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD05664009 CAS číslo: 128717-77-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...